Learn More
Desonide 21-Acetate, TRC
Product Code. 30350355
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350355 100 mg 100mg
30358744 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30350355 Supplier Toronto Research Chemicals Supplier No. TRCD296925100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25092-25-5
Molecular Formula C26 H34 O7
Molecular Weight (g/mol) 458.54
Synonym Acetyldesonide, 11β-Hydroxy-16α,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxopregna-1,4-dien-21-yl acetate, Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-acetate (7CI,8CI), (11β,16α)-21-(Acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Desonide 21-acetate
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C26H34O7/c1-14(27)31-13-20(30)26-21(32-23(2,3)33-26)11-18-17-7-6-15-10-16(28)8-9-24(15,4)22(17)19(29)12-25(18,26)5/h8-10,17-19,21-22,29H,6-7,11-13H2,1-5H3/t17-,18-,19-,21+,22+,24-,25-,26+/m0/s1
Formula Weight 458.2305
Percent Purity >95%
Chemical Name or Material Desonide 21-Acetate
Grade HPLC