missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Antibody (D33), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-17156-0.5ML
This item is not returnable.
View return policy
Description
Desmin Monoclonal specifically detects Desmin in Human, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Desmin | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with sodium azide | |
| DES | |
| Full length native protein purified from human muscle. | |
| Primary | |
| NB120-17156 stains striated as well as smooth muscle cells. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| D33 | |
| Western Blot 1:50 - 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:25 - 1:50 | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Mouse | |
| 0.5 mL | |
| 1674 | |
| Human, Rat, Chicken | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction