missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Antibody (D33), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-17156
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| Desmin | |
| Monoclonal | |
| Unconjugated | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| This antibody reacts with desmin protein of 53 kDa. Both striated as well as smooth muscle cells stain well with this antibody. The antibody shows a broad interspecific cross-reactivity, reacting with human. | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| D33 | |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| DES | |
| BALB/C mice were immunized with purified desmin from human muscle. | |
| 1 mL | |
| Cytoskeleton Markers | |
| 1674 | |
| Human, Rat, Chicken | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction