Learn More
Desmethyl Metolazone, TRC
Specifications
Specifications
| Chemical Name or Material | Desmethyl Metolazone |
| CAS | 28524-40-5 |
| Molecular Formula | C15H14ClN3O3S |
| Quantity | 25 mg |
| Synonym | 7-Chloro-1,2,3,4-tetrahydro-2-methyl-4-oxo-3-phenyl-6-quinazolinesulfonamide |
| InChI Formula | InChI=1S/C15H14ClN3O3S/c1-9-18-13-8-12(16)14(23(17,21)22)7-11(13)15(20)19(9)10-5-3-2-4-6-10/h2-9,18H,1H3,(H2,17,21,22) |
| SMILES | CC1Nc2cc(Cl)c(cc2C(=O)N1c3ccccc3)S(=O)(=O)N |
| Molecular Weight (g/mol) | 351.81 |