Learn More
Desmethyl Doxylamine Oxalate, TRC
Specifications
Specifications
| Chemical Name or Material | Desmethyl Doxylamine Oxalate |
| CAS | 2771193-12-3 |
| Molecular Formula | C16H20N2O . C2H2O4 |
| Quantity | 25 mg |
| Synonym | N,N-Dimethyl-2-[phenyl(pyridin-2-yl)methoxy]ethan-1-amine Oxalate |
| InChI Formula | InChI=1S/C16H20N2O.C2H2O4/c1-18(2)12-13-19-16(14-8-4-3-5-9-14)15-10-6-7-11-17-15;3-1(4)2(5)6/h3-11,16H,12-13H2,1-2H3;(H,3,4)(H,5,6) |
| SMILES | CN(C)CCOC(C1=CC=CC=C1)C2=NC=CC=C2.O=C(O)C(O)=O |
| IUPAC Name | N,N-dimethyl-2-(phenyl(pyridin-2-yl)methoxy)ethan-1-amine oxalate |
| Molecular Weight (g/mol) | 346.37 |
| Formula Weight | 346.16 |