Learn More
Deshydroxyethoxy-2,3-O-(dimethylmethylene) Ticagrelor, TRC
Specifications
Specifications
| Chemical Name or Material | Deshydroxyethoxy-2,3-O-(dimethylmethylene) Ticagrelor |
| CAS | 274693-49-1 |
| Molecular Formula | C24H28F2N6O3S |
| Quantity | 1 g |
| Synonym | (3aR,4S,6R,6aS)-6-[7-[[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino]-5-(propylthio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-3-yl]tetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol |
| InChI Formula | InChI=1S/C24H28F2N6O3S/c1-4-7-36-23-28-21(27-15-9-12(15)11-5-6-13(25)14(26)8-11)18-22(29-23)32(31-30-18)16-10-17(33)20-19(16)34-24(2,3)35-20/h5-6,8,12,15-17,19-20,33H,4,7,9-10H2,1-3H3,(H,27,28,29)/t12-,15+,16+,17-,19-,20+/m0/s1 |
| SMILES | CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](O)[C@H]6OC(C)(C)O[C@@H]56)c4n1 |
| IUPAC Name | (3aR,4S,6R,6aS)-6-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfanyltriazolo[4,5-d]pyrimidin-3-yl]-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-ol |
| Molecular Weight (g/mol) | 518.58 |
| Formula Weight | 518.1912 |