Learn More
Desamino P-Ethoxy Glufosinate-d3 Ethyl Ester, TRC
Product Code. 30352031
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352031 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30352031 Supplier Toronto Research Chemicals Supplier No. TRCD28845725MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Desamino P-Ethoxy Glufosinate-d3 Ethyl Ester
CAS 1346598-74-0
Molecular Formula C9H16D3O4P
Quantity 25 mg
Synonym 4-(Hydroxymethylphosphinyl)-butanoic Acid-d3
InChI Formula InChI=1S/C9H19O4P/c1-4-12-9(10)7-6-8-14(3,11)13-5-2/h4-8H2,1-3H3/i3D3
SMILES [2H]C([2H])([2H])P(=O)(CCCC(=O)OCC)OCC
IUPAC Name ethyl 4-[ethoxy(trideuteriomethyl)phosphoryl]butanoate
Molecular Weight (g/mol) 225.24
Formula Weight 225.1209