Learn More
Desacetyl Triflusal, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Trifluoromethyl)salicylic Acid |
| CAS | 328-90-5 |
| Molecular Formula | C8 H5 F3 O3 |
| Quantity | 5 g |
| Synonym | 2-Hydroxy-4-(trifluoromethyl)-benzoic Acid, 2,4-Cresotic acid, α,α,α-trifluoro- (6CI,7CI,8CI), 2-Hydroxy-4-(trifluoromethyl)benzoic acid, 4-(Trifluoromethyl)salicylic acid, 4-Trifluoromethyl-2-hydroxybenzoic acid, p-(Trifluoromethyl)salicylic acid |
| InChI Formula | InChI=1S/C8H5F3O3/c9-8(10,11)4-1-2-5(7(13)14)6(12)3-4/h1-3,12H,(H,13,14) |
| SMILES | OC(=O)c1ccc(cc1O)C(F)(F)F |
| IUPAC Name | 2-hydroxy-4-(trifluoromethyl)benzoic acid |
| Molecular Weight (g/mol) | 206.12 |
| Formula Weight | 206.0191 |