Learn More
Des-(Cyclohexa-2,5-dien-1-one) Androsta-(E)-3-oxobut-1-en-1-yl-2,3,4-13C3), TRC
Specifications
Specifications
| Chemical Name or Material | Des-(Cyclohexa-2,5-dien-1-one) Androsta-(E)-3-oxobut-1-en-1-yl-2,3,4-13C3 |
| CAS | 327049-06-9 |
| Molecular Formula | C1613C3H26O3 |
| Quantity | 1 mg |
| Synonym | Des-(Cyclohexa-2,5-dien-1-one) Boldenone-(E)-3-oxobut-1-en-1-yl-2,3,4-13C3), rac-(3aR,5aS,6S,9aS,9bS)-3a,6-Dimethyl-6-((E)-3-oxobut-1-en-1-yl-2,3,4-13C3)decahydro-3H-cyclopenta[a]naphthalene-3,7(3aH)-dione |
| InChI Formula | InChI=1S/C19H26O3/c1-12(20)8-10-18(2)15-9-11-19(3)14(5-7-17(19)22)13(15)4-6-16(18)21/h8,10,13-15H,4-7,9,11H2,1-3H3/b10-8+/t13-,14-,15-,18+,19-/m0/s1/i1+1,8+1,12+1 |
| SMILES | [13CH3][13C](=O)\[13CH]=C\[C@]1(C)[C@H]2CC[C@@]3(C)[C@@H](CCC3=O)[C@@H]2CCC1=O |
| IUPAC Name | (3aS,5aS,6S,9aS,9bS)-3a,6-dimethyl-6-[(E)-3-oxo(2,3,4-^{13}C_{3})but-1-enyl]-2,4,5,5a,8,9,9a,9b-octahydro-1H-cyclopenta[a]naphthalene-3,7-dione |
| Molecular Weight (g/mol) | 305.39 |
| Formula Weight | 305.1983 |