Learn More
Des-[2-[(Aminoiminomethyl)amino]acetyl]amino] N-(2-Cyanoethyl)-4-nitro Netropsin Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Des-[2-[((Aminoiminomethyl)amino)acetyl]amino] N-(2-Cyanoethyl)-4-nitro Netropsin Dihydrochloride |
| CAS | 3185-94-2 |
| Molecular Formula | C15H16N6O4 |
| Quantity | 10 mg |
| Synonym | N-(2-Cyanoethyl)-1-methyl-4-(1-methyl-4-nitro-1H-pyrrole-2-carboxamido)-1H-pyrrole-2-carboxamide, N-[5-[[(2-Cyanoethyl)amino]carbonyl]-1-methyl-1H-pyrrol-3-yl]-1-methyl-4-nitro-1H-pyrrole-2-carboxamide |
| InChI Formula | InChI=1S/C15H16N6O4/c1-19-8-10(6-12(19)14(22)17-5-3-4-16)18-15(23)13-7-11(21(24)25)9-20(13)2/h6-9H,3,5H2,1-2H3,(H,17,22)(H,18,23) |
| SMILES | O=C(C1=CC([N+]([O-])=O)=CN1C)NC2=CN(C)C(C(NCCC#N)=O)=C2 |
| Molecular Weight (g/mol) | 344.33 |
| Formula Weight | 344.12 |