Learn More
DEPC-treated WaterAbnova™
Catalog Number 16092038
Click to view available options
Quantity:
5 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
DEPC-treated Water is can be used in various applications in molecular biology, paticulary for use of RNA.
  • The DEPC-treated Water was prepared by 0.1 % DEPC-treatment and autoclave of highly purified water.
  • The product was confirmed to be RNase-free and DNase free.
  • Apperance is clear liquid.
Quantity 500 mL