Learn More
Thermo Scientific Chemicals Deoxycholic acid, 98.5%
Product Code. 10636904 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. 10636904
Brand: Thermo Scientific Chemicals 148145000
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Deoxycholic acid, CAS # 83-44-3, is a bile acid and blood serum metabolite. | CAS: 83-44-3 | C24H40O4 | 392.58 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Deoxycholic acid is a 5beta-cholan-24-oic acid derivative with hydroxy substitutions at carbons 3 and 12
• Deoxycholic acid’s role is to absorb fats by emulsifying and solubilizing them
Applications
• Deoxycholic acid may be used for laboratory study of submental fat
• Deoxycholic acid is capable of disrupting cell membranes
• In laboratory research applications, deoxycholic acid can be used to solubilize proteins or as a secondary standard
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
Specifications
Melting Point 172°C to 178°C
Color Pink to White
Ignition Residue 0.2 % max.
Loss on Drying 1 % max. (140°C, 4 hrs) (under vacuum < 5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Glass bottle
Quantity 500 g
Beilstein 10, IV, 1608
Merck Index 15, 2901
Specific Rotation Condition + 55.00 (20.00°C c=1,ETHANOL)
Specific Rotation +53° to +57° (20°C, 589nm) (c=1, EtOH)
IUPAC Name (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Formula Weight 392.57
Percent Purity 98.0 to 102.0% (on dried basis) (Acid)
Physical Form Powder
Chemical Name or Material Deoxycholic acid
Show More Show Less
compliance-icons
Product Identifier
  • Deoxycholic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.