Learn More
Thermo Scientific Chemicals Deoxycholic acid, 98.5-101%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Deoxycholic acid is a 5beta-cholan-24-oic acid derivative with hydroxy substitutions at carbons 3 and 12
• Deoxycholic acid’s role is to absorb fats by emulsifying and solubilizing them
Applications
• Deoxycholic acid may be used for laboratory study of submental fat
• Deoxycholic acid is capable of disrupting cell membranes
• In laboratory research applications, deoxycholic acid can be used to solubilize proteins or as a secondary standard
Specifications
Melting Point | 172°C to 178°C |
Color | Pink to White |
Ignition Residue | 0.2 % max. |
Loss on Drying | 1 % max. (140°C, 4 hrs) (under vacuum < 5 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 10, IV, 1608 |
Merck Index | 15, 2901 |
Show More |
Safety and Handling
- Deoxycholic acid
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only