missing translation for 'onlineSavingsMsg'
Learn More

Deoxy Artemisinin, TRC

Product Code. 30358083
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
Product Code. Quantity unitSize
30358083 100 mg 100mg
30355378 5 mg 5mg
30361281 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 30358083

Brand: Toronto Research Chemicals TRCD232150100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72826-63-2
Formula Weight 266.1518
InChI Formula InChI=1S/C15H22O4/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-15/h8-11,13H,4-7H2,1-3H3/t8-,9-,10+,11+,13-,14-,15-/m1/s1
Molecular Formula C15 H22 O4
Molecular Weight (g/mol) 266.33
SMILES C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23O4
Synonym 10aH-9,10b-Epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, octahydro-3,6,9-trimethyl-, (3R,3aS,6R,6aS,9S,10aS,10bR)-, 10aH-9,10b-Epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, octahydro-3,6,9-trimethyl-, [3R-(3α,3aβ,6β,6aβ,9β,10aα,10bβ)]-, (3R,3aS,6R,6aS,9S,10aS,10bR)-Octahydro-3,6,9-trimethyl-10aH-9,10b-epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, 2-Deoxyartemisinin, 2-Desoxyartemisinin, Deoxyarteannuin, Deoxyartemisinin, Deoxyqinghaosu, Desoxyartemisinin, Hydroarteannuin, Qing Hau Sau III, Qinghaosu III, (3R,3aS,3a1R,6R,6aS,9S,10aS)-3,6,9-trimethyldecahydro-2H-3a1,9-epoxyoxepino[4,3,2-ij]isochromen-2-one
Chemical Name or Material Deoxy Artemisinin

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.