Learn More
Elma™ Dental Cleaner
Product Code. 10733753
Change view
Click to view available options
Volume (Metric):
10 L
25 L
Unit Size:
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10733753 10 L 10 L
10496012 25 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10733753 Supplier Elma™ Supplier No. 5813900000
Please to purchase this item. Need a web account? Register with us today!
Specifications
Product Type EC 25 Cleaner
Certifications/Compliance CE
Container Type Can
For Use With (Application) For the thorough ultrasonic cleaning of impression trays and dental instruments made from Stainless Steel, Glass, Ceramics and Plastics.
Volume (Metric) 10 L