missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dendritic Cell Marker Rat anti-Mouse, PE, Clone: AP-MAB0801, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-85474PE-0.1ML
This item is not returnable.
View return policy
Description
Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Dendritic Cell Marker | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| Protein G purified | |
| RUO | |
| This reacts with Dendritic Cell Inhibitory Receptor 2 on mouse dendritic cells. | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry | |
| AP-MAB0801 | |
| Flow Cytometry | |
| DCs, Dendritic cell | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse dendritic cells (DCs). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction