Learn More
delta-1,2,3,4,5,6-Hexachlorocyclohexane (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | delta-HCH |
| CAS | 319-86-8 |
| Molecular Formula | C6 H6 Cl6 |
| Quantity | 5 mg |
| Synonym | (1α,2α,3α,4β,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane, Delta-BHC, Δ-1,2,3,4,5,6-Hexachlorocyclohexane, Δ-666, Δ-BHC, Δ-Benzene hexachloride, Δ-HCH, Δ-Hexachlorocyclohexane, Δ-Lindane |
| InChI Formula | InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5-,6- |
| SMILES | Cl[C@@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@H]1Cl |
| IUPAC Name | 1,2,3,4,5,6-hexachlorocyclohexane |
| Molecular Weight (g/mol) | 290.83 |
| Formula Weight | 287.8601 |