Learn More
DEL-22379, MedChemExpress
Product Code. 16443657
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16443657
Brand: Medchem Express HY18932
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress DEL-22379 is an ERK dimerization Inhibitor. DEL-22379 readily binds to ERK2 with a Kd estimated in the low micromolar range, though binding is detectable even at low nanomolar concentrations. ERK2 dimerization is progressively inhibited with an IC50 of ∼0.5 μM.
DEL-22379 is an ERK dimerization Inhibitor. DEL-22379 readily binds to ERK2 with a Kd estimated in the low micromolar range, though binding is detectable even at low nanomolar concentrations. ERK2 dimerization is progressively inhibited with an IC50 of ∼0.5 μM.
Specifications
Chemical Name or Material DEL-22379
CAS 181223-80-3
Color Orange
Purity Grade Notes Research
Molecular Formula C26H28N4O3
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (224.96 mM; Need ultrasonic)
SMILES O=C(CCN1CCCCC1)NC2=CC=C(NC(/C3=C\C4=CNC5=C4C=C(OC)C=C5)=O)C3=C2
Molecular Weight (g/mol) 444.53
Formula Weight 444.53
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease