missing translation for 'onlineSavingsMsg'
Learn More

Dehydrocorydaline, MedChemExpress

Product Code. 16441478
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16441478

Brand: Medchem Express HYN0674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dehydrocorydaline (13-Methylpalmatine) is an alkaloid that regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. Dehydrocorydaline elevates p38 MAPK activation. Anti-inflammatory and anti-cancer activities. Dehydrocorydaline shows strong anti-malarial effects (IC50=38 nM), and low cytotoxicity (cell viability > 90%) using P. falciparum 3D7 strain.

Dehydrocorydaline (13-Methylpalmatine) is an alkaloid that regulates protein expression of Bax, Bcl-2

Specifications

Chemical Name or Material Dehydrocorydaline
Physical Form Solid
CAS 30045-16-0
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H24NO4+
For Use With (Application) Neuroscience-Neurodegeneration
Synonym 13-Methylpalmatine
Solubility Information DMSO : 50 mg/mL (136.45 mM; Need ultrasonic and warming)
SMILES CC1=C(C=CC(OC)=C2OC)C2=C[N+]3=C1C4=CC(OC)=C(OC)C=C4CC3
Molecular Weight (g/mol) 366.43
Formula Weight 366.43
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.