Learn More
Dehydro Aripiprazole-d8, TRC
Specifications
Specifications
| Chemical Name or Material | Dehydro Aripiprazole-d8 |
| CAS | 2749328-53-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23H17D8Cl2N3O2 |
| Quantity | 1 mg |
| Synonym | 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy-d8]-2(1H)-quinolinone |
| InChI Formula | InChI=1S/C23H25Cl2N3O2.ClH/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18;/h3-9,16H,1-2,10-15H2,(H,26,29);1H/i11D2,12D2,13D2,14D2; |
| SMILES | ClC1=CC=CC(N2CCN(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OC3=CC=C(C=CC(N4)=O)C4=C3)CC2)=C1Cl |
| Molecular Weight (g/mol) | 454.42 |
| Percent Purity | >95 |