missing translation for 'onlineSavingsMsg'
Learn More

Degrasyn, MedChemExpress

Product Code. 16405608
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405608

Brand: Medchem Express HY13264

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Degrasyn (WP1130) is a cell-permeable deubiquitinase (DUB) inhibitor, directly inhibiting DUB activity of USP9x, USP5, USP14, and UCH37. Degrasyn has been shown to downregulate the antiapoptotic proteins Bcr-Abl and JAK2.

Degrasyn (WP1130) is a cell-permeable deubiquitinase (DUB) inhibitor, directly inhibiting DUB activity of USP9x, USP5, USP14, and UCH37. Degrasyn has been shown to downregulate the antiapoptotic proteins Bcr-Abl and JAK2.

Specifications

Chemical Name or Material Degrasyn
CAS 856243-80-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H18BrN3O
Quantity 10 mg
Synonym WP1130
Solubility Information DMSO : 100 mg/mL (260.23 mM; Need ultrasonic)
SMILES BrC1=CC=CC(/C=C(C(N[C@H](C2=CC=CC=C2)CCC)=O)\C#N)=N1
Molecular Weight (g/mol) 384.27
Formula Weight 384.27
Percent Purity 93.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.