missing translation for 'onlineSavingsMsg'
Learn More

Deferasirox, MedChemExpress

Product Code. 16427256
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427256 100 mg 100mg
16483566 25 mg 25mg
16479807 5 mg 5mg
16453317 50 mg 50mg
16417308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16427256 Supplier Medchem Express Supplier No. HY17359

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Deferasirox (ICL 670) is an orally available iron chelator used for the management of transfusional iron overload.

Deferasirox (ICL 670) is an orally available iron chelator used for the management of transfusional iron overload.

Specifications

Chemical Name or Material Deferasirox
CAS 201530-41-8
Color White
Purity Grade Notes Research
Molecular Formula C21H15N3O4
Quantity 100 mg
Synonym ICL 670
Solubility Information DMSO : ≥ 100 mg/mL (267.84 mM)
SMILES O=C(O)C1=CC=C(N2N=C(C3=CC=CC=C3O)N=C2C4=CC=CC=C4O)C=C1
Molecular Weight (g/mol) 373.36
Formula Weight 373.36
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.