Learn More
Defensin 6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553992
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PLQAEDDPLQ AKAYEADAQE QRGANDQDFA VSFAEDASSS LRALGSTRAF TCHCRRSCYS TEYSYGTCTV MGINHRFC Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 35%.
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8.Specifications
Defensin 6 | |
Polyclonal | |
Unconjugated | |
DEFA6 | |
DEF6; DEFA6; defensin 6; defensin alpha 6; Defensin, alpha 6; defensin, alpha 6, Paneth cell-specific; defensin-6; HD-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1671 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q01524 | |
DEFA6 | |
Recombinant protein corresponding to Human Defensin 6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only