missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEFB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14738-1-AP-150UL
This item is not returnable.
View return policy
Description
Beta-defensins (also designated BD, and hBD in human) are small cationic peptides with broad-spectrum antimicrobial activity. Produced in mucosal epithelia and neutrophils of several species, beta-defensins are developmentally regulated. Research indicates that human beta-defensin 1 may contribute to the antimicrobial activity of airway surface fluid and may play a role in the mucosal defenses of the lung. Human beta-defensin 1 shares homology with other beta-defensins from human blood filtrate and is also present in omolar concentrations in human plasma. In addition to the antimicrobial activity of human airway epithelia, beta-defensin 1 may play a role in the mucosal defenses of the lung.Specifications
| DEFB1 | |
| Polyclonal | |
| Unconjugated | |
| DEFB1 | |
| BD 1, BD1, Beta defensin 1, DEFB 1, DEFB1, DEFB101, defensin, beta 1, hBD 1, HBD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1672 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P60022 | |
| DEFB1 | |
| DEFB1 Fusion Protein Ag6406 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction