Learn More
BM Polyco™ Deep Sink™ Gloves
Click to view available options
Size:
7
8
Unit Size:
Pack of 120
Description
- Medium-weight glove offering improved strength with a high level of dexterity.
- Slip-resistant pattern on the palm and fingers optimizes grip especially in wet conditions.
- Anatomically shaped and flexible to minimize hand fatigue.
- Soft cotton flock lining absorbs perspiration to keep hands cool, dry and comfortable.
- Extra-long cuff provides extra protection to the forearm.
- Beaded cuff adds strength and prevents liquid rollback.
- Ideal for use as a general handling janitorial glove.
Specifications
Specifications
| Material | Latex |
| Size | 7 |
| Length (Metric) | 40 cm |