Learn More
BM Polyco™ Deep Sink™ Gloves
Product Code. p-8009687
Click to view available options
Size:
7
8
Unit Size:
Pack of 120
Product Code. 15364720
Brand: BM Polyco™ 625
Only null left
Add to basket
Add to basket
BM Polyco™ Deep Sinks™ Gloves are extra-long, medium-weight, latex gloves with a soft cotton flock lining.
  • Medium-weight glove offering improved strength with a high level of dexterity.
  • Slip-resistant pattern on the palm and fingers optimizes grip especially in wet conditions.
  • Anatomically shaped and flexible to minimize hand fatigue.
  • Soft cotton flock lining absorbs perspiration to keep hands cool, dry and comfortable.
  • Extra-long cuff provides extra protection to the forearm.
  • Beaded cuff adds strength and prevents liquid rollback.
  • Ideal for use as a general handling janitorial glove.
Specifications
Material Latex
Size 7
Length (Metric) 40 cm