Learn More
Borer Chemie™ deconex 21 LAB CLF
Description
- Concentrate for automated cleaning
- High yield, typical concentration: 5-10 mL/L
- Excellent material compatibility, also for aluminium
- Broad cleaning spectrum
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Without chlorine, EDTA, NTA and surfactants
- Acid rinse with deconex cleaner recommended
- Technical support by the manufacturer
Specifications
Specifications
| Form | Liquid |
| pH | ∼11.4 (1% Solution) |
| Composition | Alkalis, complexing agents, corrosion inhibitor, sequestering agents |
| Concentration | Concentrate |
| Container Type | Canister |
| Content And Storage | 5°C to 25°C |
| Quantity | 1 x 32 kg |
| Volume (English) | 6.2 gal. |
| Volume (Metric) | 23.5 L |
| Weight (English) | 70.5 lb. |
| Show More |