Learn More
Decloxizine dihydrochloride, MedChemExpress
Description
Decloxizine dihydrochloride(UCB-1402
Specifications
Specifications
| Chemical Name or Material | Decloxizine dihydrochloride |
| Physical Form | Solid |
| CAS | 13073-96-6 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H30Cl2N2O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | UCB 1402 dihydrochloride |
| Solubility Information | H2O : ≥ 100 mg/mL (241.91 mM) ∣DMSO : ≥ 50 mg/mL (120.95 mM) |
| Show More |