Learn More
Deacetyl Ketoconazole, TRC
Product Code. 30137528
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137528 500 mg 500mg
30134053 250 mg 250mg
18031350 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137528 Supplier Toronto Research Chemicals Supplier No. TRCD198955500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67914-61-8
Molecular Formula C24 H26 Cl2 N4 O3
Molecular Weight (g/mol) 489.39
Synonym 1-[4-[[(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, Ketoconazole Imp. D (EP)
IUPAC Name 1-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
SMILES Clc1ccc(c(Cl)c1)[C@@]2(Cn3ccnc3)OC[C@H](COc4ccc(cc4)N5CCNCC5)O2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C24H26Cl2N4O3/c25-18-1-6-22(23(26)13-18)24(16-29-10-7-28-17-29)32-15-21(33-24)14-31-20-4-2-19(3-5-20)30-11-8-27-9-12-30/h1-7,10,13,17,21,27H,8-9,11-12,14-16H2/t21-,24-/m0/s1
Formula Weight 488.1382
Percent Purity >95%
Chemical Name or Material Deacetyl Ketoconazole
Grade HPLC