missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX5 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98889FR
This item is not returnable.
View return policy
Description
DDX5 Polyclonal specifically detects DDX5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DDX5 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 1655 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 680 | |
| B-cell CLL/lymphoma 7 protein family member C, B-cell CLL/lymphoma 7C | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human DDX5. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction