missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DDX47 Polyclonal antibody specifically detects DDX47 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | DDX47 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | DEAD (Asp-Glu-Ala-Asp) box polypeptide 47, DEAD box polypeptide 47, DEAD box protein 47, DKFZp564O176, E4-DBP, E4-DEAD box protein, EC 3.6.1, EC 3.6.4.13, FLJ30012, HQ0256, MSTP162, probable ATP-dependent RNA helicase DDX47, RRP3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 186-455 of human DDX47 (NP_057439.2).,, Sequence:, TEVDKILKVIPRDRKTFLFSATMTKKVQKLQRAALKNPVKCAVSSKYQTVEKLQQYYIFIPSKFKDTYLVYILNELAGNSFMIFCSTCNNTQRTALLLRNLGFTAIPLHGQMSQSKRLGSLNKFKAKARSILLATDVASRGLDIPHVDVVVNFDIPTHSKDYIHRVGRTARAGRSGKAITFVTQYDVELFQRIEHLIGKKLPGFPTQDDEVMMLTERVAEAQRFARMELREHGEKKKRSREDAGDNDDTEGAIGVRNKVAGGKMKKRKGR |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?