Learn More
DDX26B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553044
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DNLDCGLSYS VISYLKKLSQ QTKLESERIL ASVGKKPPQE IGIKVKNHSG GGMSLTHNKN FRKLLKEITG ETALRLTELN TKEFAGFQIG LLNKDLKPQT YRNAYDIPRR GLLDQLTRMR SNLLKTH Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 76%.
DDX26B is a protein coding gene. An important paralog of this gene is INTS6.Specifications
DDX26B | |
Polyclonal | |
Unconjugated | |
INTS6L | |
4930535D10Rik; 6330505F04Rik; D130066O12; ddx26b; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B; hypothetical protein MGC63527; int6; Integrator complex subunit 6; integrator complex subunit 6 like; integrator complex subunit 6-like; ints6; Ints6l; LOW QUALITY PROTEIN: integrator complex subunit 6-like; protein DDX26B; zgc:63527 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
203522 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JSJ4 | |
INTS6L | |
Recombinant protein corresponding to Human DDX26B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only