missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDR2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA587602

Product Code. 16586051

  • 520.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: NMFAKGVKIF KEVQCYFRSE ASEWEPNAIS FPLVLDDVNP SARFVTVPLH HRMASAIKCQ YHFADTWMMF SEITFQSDAA MYNNSEALPT SPMAPTTYDP MLKVDDSNTR I; Positive Samples: HeLa, A375, HT-1080, Mouse lung, Mouse heart, Mouse kidney, Rat lung; Cellular Location: Cell membrane, Single-pass type I membrane protein.

DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain. DDR1 and DDR2 have been shown to be potently inhibited by Gleevec. DDR2 required for normal bone development. Regulates osteoblast differentiation and chondrocyte maturation via a signaling pathway that involves MAP kinases and leads to the activation of the transcription factor RUNX2. Regulates remodeling of the extracellular matrix by up-regulation of the collagenases MMP1, MMP2 and MMP13, and thereby facilitates cell migration and tumor cell invasion. Promotes fibroblast migration and proliferation, and thereby contributes to cutaneous wound healing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDR2
Polyclonal
Unconjugated
Ddr2
AW495251; CD_antigen: CD167b; CD167 antigen-like family member B; CD167b; CD167b antigen; cell migration-inducing protein 20; Ddr2; Discoidin domain receptor 2; discoidin domain receptor family, member 2; discoidin domain receptor tyrosine kinase 2; discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein; hydroxyaryl-protein kinase; MIG20a; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; neurotrophic tyrosine kinase, receptor-related 3; Ntrkr3; receptor protein-tyrosine kinase TKT; RP11-572K18.1; TKT; TYRO10; tyrosine-protein kinase TYRO 10; tyrosine-protein kinase TYRO10; tyrosylprotein kinase
Rabbit
Affinity chromatography
RUO
18214, 4921, 685781
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16832, Q62371
Ddr2
Recombinant fusion protein containing a sequence corresponding to amino acids 290-400 of human DDR2 (NP_0010147961).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.