missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCTN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16947-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains an RGD (Arg-Gly-Asp) motif in the N-terminal region, which confers adhesive properties to macromolecular proteins like fibronectin. It shares a high degree of sequence similarity with the mouse homolog, which has been suggested to play a role in mitochondrial biogenesis. The exact biological function of this gene is not known.Specifications
| DCTN6 | |
| Polyclonal | |
| Unconjugated | |
| DCTN6 | |
| DCTN6, dynactin 6, Dynactin subunit 6, Dynactin subunit p27, Protein WS 3, WS 3, WS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10671, 22428, 290798 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00399, Q9WUB4 | |
| Dctn6 | |
| DCTN6 Fusion Protein Ag10561 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction