Learn More
DCT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552741
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NECDVCTDQL FGAARPDDPT LISRNSRFSS WETVCDSLDD YNHLVTLCNG TYEGLLRRNQ MGRNSMKLPT LKDIRDCLSL QKFDNPPFFQ NSTFSFRNAL EGFDKADGTL DSQVMSLHNL VHSFLNGTNA LPHSAANDPI FVVLHSFTDA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 52%.
The TYR-related proteins, TRP1 and TRP2, are also specifically expressed in melanocytes, and they likewise contribute to the synthesis of melanin within the melanosomes. The TRPs, including TYR, all share a similar transmembrane region, contain two metal-binding regions and a cysteinerich epidermal growth factor motif, and are localized in the melanosomal membrane. These proteins, however, have distinct catalytic activity, and they individually contribute to the biosynthesis of melanin biopolymers. The TRPs are believed to exist as a multi-enzyme complex, as these proteins form aggregates together, and the expression of TRP1 also helps stabilize TYR in melanocytes.Specifications
DCT | |
Polyclonal | |
Unconjugated | |
DCT | |
Dct; DOPAchrome conversion factor; dopachrome delta isomerase; dopachrome delta-isomerase; DOPAchrome isomerase; DOPAchrome oxidoreductase; dopachrome tautomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; hypothetical protein LOC664745; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; melanogenic enzyme; tyrosinase-related protein 2; trp2; RGD1564975; slaty; SLATY locus protein; slt; trp2; TRP-2; tyrosinase related protein 2; tyrosinase related protein-2; tyrosinase-related protein 2; tyrosinase-related protein-2; tyrosine-related protein 2; Tyrp2; Tyrp-2; zgc:109860 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1638 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P40126 | |
DCT | |
Recombinant protein corresponding to Human DCT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only