missing translation for 'onlineSavingsMsg'
Learn More

dBET6, MedChemExpress

Product Code. 16434658
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434658

Brand: Medchem Express HY112588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress dBET6 is a highly potent, selective and cell-permeable PROTAC connected by ligands for Cereblon and BET, with an IC50 of 14 nM, and has antitumor activity.

dBET6 is a highly potent, selective and cell-permeable PROTAC connected by ligands for Cereblon and BET, with an IC50 of 14 nM, and has antitumor activity.

Specifications

Chemical Name or Material dBET6
CAS 1950634-92-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C42H45ClN8O7S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (59.43 mM; Need ultrasonic)
SMILES O=C(NCCCCCCCCNC(COC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)=O)C[C@H]4C5=NN=C(C)N5C6=C(C(C)=C(C)S6)C(C7=CC=C(Cl)C=C7)=N4
Molecular Weight (g/mol) 841.37
Formula Weight 841.37
Percent Purity 98.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.