missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DBCO-(PEG)3-VC-PAB-MMAE

Product Code. 16451149
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451149 1 mg 1mg
16437117 5 mg 5mg
16414398 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451149 Supplier MedChemExpress Supplier No. HY111012
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DBCO-(PEG)3-VC-PAB-MMAE is made by MMAE conjugated to DBCO-(PEG)3-vc-PAB linker. Monomethyl auristatin E (MMAE), a potent tubulin inhibitor, is a toxin payload in antibody drug conjugate.

Specifications

Molecular Weight (g/mol) 1613.97 g/mol
Content And Storage -20°C, stored under nitrogen ∣ The compound is unstable in solutions, freshly prepared is recommended.
Shipping Condition Room temperature
Quantity 1 mg
Solubility DMSO : 60 mg/mL (37.18 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.