Learn More
Agilent Technologies DB-XLB 30 m, 0.25mm, 0.25um, 10m DRgrD
Brand:  Agilent Technologies 122-1232G
Encompass_Preferred
DB-XLB 30 m, 0.25mm, 0.25um, 10m DRgrD
Product Code. 10080963
  • 1194.27 € / Each
Only null left
Estimated Shipment: 16-01-2026