Learn More
Agilent Technologies DB-WAXETR 60m, 0,32mm, 0,50um,5' Korb
Brand:  Agilent Technologies 123-7363E
Encompass_Preferred
DB-WAXETR 60m, 0,32mm, 0,50um,5' Korb
Product Code. 10004783
  • 1945.68 € / Each
Only null left
Estimated Shipment: 21-01-2026