Learn More
Agilent Technologies DB-5 30M 0.32mm 0.25U
Brand:  Agilent Technologies 123-5032
Encompass_Preferred
DB-5 30M 0.32mm 0.25U
Product Code. 10011693
  • 981.31 € / Each
Only null left
Estimated Shipment: 22-01-2026