Learn More
Agilent Technologies DB-17 30M 0.25mm 0.25U
Brand:  Agilent Technologies 122-1732
Encompass_Preferred
DB-17 30M 0.25mm 0.25U
Product Code. 10051683
  • 908.71 € / Each
Only null left
Estimated Shipment: 21-01-2026