Learn More
Agilent Technologies DB-1301 30M 0.25mm 1.00U
Brand:  Agilent Technologies 122-1333
Encompass_Preferred
DB-1301 30M 0.25mm 1.00U
Product Code. 10000543
  • 943.80 € / Each
Only null left
Estimated Shipment: 21-01-2026