Learn More
Emma Safety Footwear Daytona Safety Shoes
Click to view available options
Size:
35
36
40
45
46
47
48
Unit Size:
1 pair
Description
- S1-Safety shoe for dry working conditions
- Nubuck construction
- GripForce™ Easy Twist Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Velcro closure
- Sanitized
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Velcro |
| Color | Black, Gray |
| Design | Daytona |
| Liner Material | Hydro-Tec™ Sanitized Silver |
| Material | Nubuck |
| Size | 35 |
| Toe Material | Steel |