missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP12 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52376
This item is not returnable.
View return policy
Description
DAP12 Polyclonal antibody specifically detects DAP12 in Human, Bat, Canine, Equine, Primate, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Specifications
| DAP12 | |
| Polyclonal | |
| Unconjugated | |
| TBS (pH 7.3) and 0.5% BSA | |
| DAP12PLOSL, DNAX-activation protein 12, KARAPPLO-SL, KAR-associated protein, killer activating receptor associated protein, Killer-activating receptor-associated protein, TYRO protein tyrosine kinase binding protein, TYRO protein tyrosine kinase-binding protein | |
| Synthetic peptide C-KQRITETESPYQE from an internal region of human DAP12 (NP_003323.1; NP_937758.1). | |
| 0.05 mg | |
| Apoptosis, Signal Transduction | |
| 7305 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 3-6 μg/mL, Immunohistochemistry-Paraffin 3-6 μg/mL, Peptide ELISA | |
| O43914 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Bat, Canine, Equine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction