Learn More
Invitrogen™ Dansyl Ethylenediamine, 5-Dimethylaminonaphthalene-1-(N-(2-Aminoethyl))sulfonamide
Description
The primary aliphatic amine of dansyl ethylenediamine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). This fluorophore is lipophilic and environmentally-sensitive.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | Dansyl Ethylenediamine |
| Content And Storage | Store at room temperature and protect from light. |
| Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Amine, Ethylenediamine |
| Label or Dye | Dansyl |
| Label Type | Classic Dyes |