missing translation for 'onlineSavingsMsg'
Learn More

Daidzein 4’-Sulfate Disodium Salt, TRC

Product Code. 30358017
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358017 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358017 Supplier Toronto Research Chemicals Supplier No. TRCD10352010MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Daidzein 4'-Sulfate Disodium Salt
Molecular Formula C15H8Na2O7S
Quantity 10 mg
Synonym Sulfuric Acid Mono-[4-(7-hydroxy-4-oxo-4H-chromen-3-yl)-phenyl]ester Disodium Salt, 7-Hydroxy-3-[4-(sulfooxy)phenyl]-4H-1-benzopyran-4-one Disodium Salt, Daidzein-4'-O-sulfate Disodium Salt
SMILES O=C1C2=CC=C(O[Na])C=C2OC=C1C3=CC=C(OS(=O)(O[Na])=O)C=C3
Molecular Weight (g/mol) 378.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.