missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dactolisib Tosylate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
Dactolisib Tosylate (BEZ235 Tosylate) is a dual PI3K and mTOR kinase inhibitor with IC50 values of 4, 75, 7, 5 nM for PI3Kα, β, γ, δ, respectively. Dactolisib Tosylate (BEZ235 Tosylate) inhibits mTORC1 and mTORC2.
Specifications
Specifications
| Chemical Name or Material | Dactolisib Tosylate |
| CAS | 1028385-32-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C37H31N5O4S |
| Quantity | 500 mg |
| Synonym | BEZ235 TosylateNVP-BEZ 235 Tosylate |
| Solubility Information | DMSO : 34 mg/mL (52.98 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | CN(C1=C2C3=CC(C4=CC5=CC=CC=C5N=C4)=CC=C3N=C1)C(N2C6=CC=C(C=C6)C(C)(C#N)C)=O.CC7=CC=C(S(=O)(O)=O)C=C7 |
| Molecular Weight (g/mol) | 641.74 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction