missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DACT3/Dapper 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03293-100UL
This item is not returnable.
View return policy
Description
DACT3/Dapper 3 Polyclonal specifically detects DACT3/Dapper 3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| DACT3/Dapper 3 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 147906 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Antagonist of beta-catenin Dapper homolog 3, arginine rich region 1, Arginine-rich region 1 protein, dapper homolog 3, dapper, antagonist of beta-catenin, homolog 3 (Xenopus laevis), MGC15476, RRR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human DACT3/Dapper 3 (NP_659493.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction