missing translation for 'onlineSavingsMsg'
Learn More

Dacomitinib, MedChemExpress

Product Code. p-200098265
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. Quantity unitSize
16496146 100 mg 100mg
16485608 10 mg 10mg
16471777 50 mg 50mg
16438307 5 mg 5mg
16421076 200 mg 200mg
5 options
This item is not returnable. View return policy

Product Code. 16496146

Brand: Medchem Express HY13272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dacomitinib (PF-00299804) is a specific and irreversible inhibitor of the ERBB family of kinases with IC50s of 6 nM, 45.7 nM and 73.7 nM for EGFR, ERBB2, and ERBB4, respectively.

Dacomitinib (PF-00299804) is a specific and irreversible inhibitor of the ERBB family of kinases with IC50s of 6 nM, 45.7 nM and 73.7 nM for EGFR, ERBB2, and ERBB4, respectively.

Specifications

Chemical Name or Material Dacomitinib
CAS 1110813-31-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H25ClFN5O2
Quantity 100 mg
Synonym PF-00299804 PF-299804
Solubility Information DMSO : 40 mg/mL (85.12 mM; Need ultrasonic)
SMILES O=C(/C=C/CN1CCCCC1)NC2=CC3=C(C=C2OC)N=CN=C3NC4=CC=C(C(Cl)=C4)F
Molecular Weight (g/mol) 469.94
Formula Weight 469.94
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.