Learn More
Daclatasvir dihydrochloride, MedChemExpress
Product Code. 16473988
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16473988
Brand: Medchem Express HY10465
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Daclatasvir dihydrochloride (BMS-790052 dihydrochloride) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir dihydrochloride is also an organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Daclatasvir dihydrochloride (BMS-790052 dihydrochloride) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir dihydrochloride is also an organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Specifications
Chemical Name or Material Daclatasvir dihydrochloride
CAS 1009119-65-6
Color White
Purity Grade Notes Research
Molecular Formula C40H52Cl2N8O6
Quantity 10 mg
Synonym BMS-790052 dihydrochlorideEBP 883 dihydrochloride
Solubility Information DMSO : ≥ 56 mg/mL (68.98 mM) ∣H2O : 50 mg/mL (61.59 mM; Need ultrasonic)
SMILES O=C(N1CCC[C@H]1C2=NC=C(C3=CC=C(C4=CC=C(C=C4)C5=CN=C(N5)[C@@H]6CCCN6C([C@H](C(C)C)NC(OC)=O)=O)C=C3)N2)[C@H](C(C)C)NC(OC)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 811.8
Formula Weight 811.8
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus