Learn More
Daclatasvir dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Daclatasvir dihydrochloride (BMS-790052 dihydrochloride) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir dihydrochloride is also an organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Daclatasvir dihydrochloride |
| CAS | 1009119-65-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C40H52Cl2N8O6 |
| Quantity | 50 mg |
| Synonym | BMS-790052 dihydrochlorideEBP 883 dihydrochloride |
| Solubility Information | DMSO : ≥ 56 mg/mL (68.98 mM) ∣H2O : 50 mg/mL (61.59 mM; Need ultrasonic) |
| SMILES | O=C(N1CCC[C@H]1C2=NC=C(C3=CC=C(C4=CC=C(C=C4)C5=CN=C(N5)[C@@H]6CCCN6C([C@H](C(C)C)NC(OC)=O)=O)C=C3)N2)[C@H](C(C)C)NC(OC)=O.[H]Cl.[H]Cl |
| Molecular Weight (g/mol) | 811.8 |
| Show More |